
2-Bromo-2'-fluoro-4'-methoxyacetophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:157014-35-2
:
Formula:C₉H₈BrFO₂
MDL Number:MFCD03730100
:247.0600 g/mol
Melting Pt:63…66 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together