Order Entry
Czech Republic
ContactUsLinkComponent
Antimony(V) fluoride compound with graphite (50:50%)
Corrosive
Toxic
Antimony(V) fluoride compound with graphite (50:50%)
Catalog # APOSPC1131-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Toxic
Antimony(V) fluoride compound with graphite (50:50%)
Catalog # APOSPC1131-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :228.75 g/mol
Storage Temperature:Ambient
UN:2923
ADR:8+6.1,II
 :GHS05
 :GHS06
CAS Number:59839-60-0
Formula:C:SbF₅
MDL Number:MFCD00236643