Methyl-2-[(2,3,4,5,6-pentafluorophenoxy)sulphonyl]benzenecarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886361-14-4
:
Formula:C₁₄H₇F₅O₅S
MDL Number:MFCD05975143
:382.26 g/mol
Melting Pt:59…61 °C
Storage Temperature:Ambient
Frequently Bought Together