
4-{3-[4-(Trifluoromethyl)phenyl]-1H-pyrazol-4-yl}pyridine 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:396129-66-1
:
Formula:C₁₅H₁₀F₃N₃
MDL Number:MFCD00800784
:289.26 g/mol
Melting Pt:>230 °C (dec.)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together