
4-{[3-(Trifluoromethyl)phenyl]sulphonyl}piperidine hydrochloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:849035-98-9
:
Formula:C₁₂H₁₅ClF₃NO₂S
MDL Number:MFCD04039237
Melting Pt:226…229 °C
:329.77 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together