
4'-(Trifluoromethyl)biphenyl-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:199528-28-4
:
Formula:C₁₄H₉F₃O₂
MDL Number:MFCD03424604
:266.22 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together