Order Entry
Czech Republic
ContactUsLinkComponent
3'-(Trifluoromethyl)[1,1'-biphenyl]-2-amine 96%
Irritant
3'-(Trifluoromethyl)[1,1'-biphenyl]-2-amine 96%
Catalog # APOSPC11163-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3'-(Trifluoromethyl)[1,1'-biphenyl]-2-amine 96%
Catalog # APOSPC11163-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :237.23 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:365-06-0
Formula:C₁₃H₁₀F₃N
MDL Number:MFCD03424654