Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium hexafluorosilicate 98%
Toxic
Ammonium hexafluorosilicate 98%
Catalog # APOSPC1103-250G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
Ammonium hexafluorosilicate 98%
Catalog # APOSPC1103-250G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
Density:2.011 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:240-968-3
ADR:6.1,III
 :178.15 g/mol
UN:2854
 :GHS06
CAS Number:16919-19-0
Formula:H₈F₆N₂Si
MDL Number:MFCD00010887