
Ammonium hexafluorophosphate 99,5%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16941-11-0
:
Formula:H₄F₆NP
MDL Number:MFCD00064642
:163 g/mol
Melting Pt:198 °C (dec.)
Density:2.18 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:241-009-1
UN:3260
ADR:8,II
:GHS05
Frequently Bought Together