
2-(2-Methoxyphenoxy)-5-(trifluoromethyl)aniline 97%
Catalog # APOSPC1079D-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:283.25 g/mol
Melting Pt:28 °C
:GHS07
Storage Temperature:Ambient
CAS Number:175135-08-7
Formula:C₁₄H₁₂F₃NO₂
MDL Number:MFCD00052419