
2,4-Dichloro-6-(trifluoromethyl)pyrimidine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16097-64-6
:
Formula:C₅HCl₂F₃N₂
MDL Number:MFCD09910264
:216.97 g/mol
Melting Pt:–48…–46 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together