Order Entry
Czech Republic
ContactUsLinkComponent
2-[(4-Fluorophenyl)sulphanyl]-5-nitrobenzenecarbaldehyde 95+%
2-[(4-Fluorophenyl)sulphanyl]-5-nitrobenzenecarbaldehyde 95+%
Catalog # APOSPC10577-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[(4-Fluorophenyl)sulphanyl]-5-nitrobenzenecarbaldehyde 95+%
Catalog # APOSPC10577-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :277.28 g/mol
Storage Temperature:Ambient
CAS Number:35985-70-7
Formula:C₁₃H₈FNO₃S
MDL Number:MFCD07021301