Order Entry
Czech Republic
ContactUsLinkComponent
1-(2,6-Difluorophenyl)-2-thiourea
Irritant
1-(2,6-Difluorophenyl)-2-thiourea
Catalog # APOSPC10574-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(2,6-Difluorophenyl)-2-thiourea
Catalog # APOSPC10574-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :188.2 g/mol
Melting Pt:151…153.5 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:59772-31-5
Formula:(C₆H₃F₂)NC(S)NH₂
MDL Number:MFCD05022144