Pentafluorophenyl-3,4-dichlorobenzenesulphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:885949-53-1
:
Formula:C₁₂H₃Cl₂F₅O₃S
MDL Number:MFCD06659538
Melting Pt:76…78 °C
Storage Temperature:Ambient
Frequently Bought Together