1-{3-Fluoro-4-[(3-methylbenzyl)oxy]phenyl}-1-ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:885949-77-9
:
Formula:C₁₆H₁₅FO₂
MDL Number:MFCD06659567
Melting Pt:68…70 °C
:258.3 g/mol
Storage Temperature:Ambient
Frequently Bought Together