1-{3-Fluoro-4-[(4-nitrobenzyl)oxy]phenyl}-1-ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:885949-79-1
:
Formula:C₁₅H₁₂FNO₄
MDL Number:MFCD06659573
:289.27 g/mol
Melting Pt:148…150 °C
Storage Temperature:Ambient
Frequently Bought Together