N'-Hydroxy-4-{[5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarboximidamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:263161-34-8
:
Formula:C₁₃H₁₀F₃N₃O₂
MDL Number:MFCD00663373
:297.24 g/mol
Melting Pt:132…133 °C
Storage Temperature:Ambient
Frequently Bought Together