
3-(3,5-Difluorophenyl)-1H-pyrazole-4-carboxaldehyde 98%
Catalog # APOSPC10358-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:208.16 g/mol
Melting Pt:178…181 °C
:GHS07
Storage Temperature:Ambient
CAS Number:875664-59-8
Formula:C₁₀H₆F₂N₂O
MDL Number:MFCD06797905