Order Entry
Czech Republic
ContactUsLinkComponent
5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-ol 95%
Irritant
5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-ol 95%
Catalog # APOSPC10340-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-ol 95%
Catalog # APOSPC10340-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :282.15 g/mol
Melting Pt:185 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:51420-73-6
Formula:C₁₀H₄F₆N₂O
MDL Number:MFCD00140206