3-(Boc-amino)-3-(4-fluorophenyl)propionic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:284493-72-7
:
Formula:C₁₄H₁₈FNO₄
MDL Number:MFCD02090715
:283.30 g/mol
Melting Pt:138…140 °C
Storage Temperature:Ambient
Frequently Bought Together