
3-Amino-3-(3-fluorophenyl)propionic acid
Catalog # APOSPC10275-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:183.1796 g/mol
Melting Pt:210…212 °C
:GHS07
Storage Temperature:Ambient
CAS Number:117391-51-2
Formula:C₉H₁₀FNO₂
MDL Number:MFCD00764457