
Ethyl-1-(4-fluorophenyl)-5-iodo-1H-pyrazole-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:959576-71-7
:
Formula:C₁₂H₁₀FIN₂O₂
MDL Number:MFCD03791218
:360.13 g/mol
Melting Pt:114…116 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together