1-Methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carbohydrazide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886360-63-0
:
Formula:C₈H₇F₃N₄OS
MDL Number:MFCD04973722
:264.23 g/mol
Melting Pt:218…220 °C
Storage Temperature:Ambient
Frequently Bought Together