
7-(Diethylamino)-2-oxo-4-(trifluoromethyl)-2H-chromene-3-carboxylic acid
Catalog # APOSPC10220-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:329.28 g/mol
Melting Pt:180…182 °C
:GHS07
Storage Temperature:Ambient
CAS Number:333303-14-3
Formula:C₁₅H₁₄F₃NO₄
MDL Number:MFCD03425755