7-Amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:863305-82-2
:
Formula:C₁₅H₁₁F₃N₄O₂
MDL Number:MFCD03791193
:336.28 g/mol
Melting Pt:224…226 °C
Storage Temperature:Ambient
Frequently Bought Together