
2,4-Bis(trifluoromethyl)imidazo[1,2-a][1,8]naphthyridine-8-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:439094-96-9
:
Formula:C₁₃H₅F₆N₃O₂
MDL Number:MFCD03425718
:349.19 g/mol
Melting Pt:281…283 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together