
Methyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}benzenecarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:385383-52-8
:
Formula:C₁₄H₉ClF₃NO₂S
MDL Number:MFCD03001227
Melting Pt:101…102 °C
:347.75 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together