
1-(6-Chloro-2-pyridinyl)-3-methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:446276-01-3
:
Formula:C₁₆H₁₂ClF₃N₄
MDL Number:MFCD03791148
:352.75 g/mol
Melting Pt:92…95 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together