
3-Methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:851975-08-1
:
Formula:C₁₁H₁₀F₃N₃
MDL Number:MFCD03791147
:241.22 g/mol
Melting Pt:110…113 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together