
Hexafluoro-2,3-bis(trifluoromethyl)-2,3-butanediol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:918-21-8
:
Formula:C₆H₂F₁₂O₂
MDL Number:MFCD00427700
:334.06 g/mol
Boiling Pt:129 °C
Melting Pt:20…24 °C
Density:1.87 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2811
ADR:6.1,II
:GHS06
Frequently Bought Together