

2-Fluoro-6-methoxybenzyl bromide
Catalog # APOSPC0899-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:219.05 g/mol
Melting Pt:18…21 °C
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
:GHS05
:GHS07
CAS Number:500912-17-4
Formula:C₈H₈BrFO
MDL Number:MFCD08688579