
4-(Pentafluorothio)benzaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:401892-84-0
:
Formula:C₇H₅F₅OS
MDL Number:MFCD03788518
:232.17 g/mol
Storage Temperature:Ambient
UN:2810
ADR:6.1,II
:GHS06
Frequently Bought Together