Ethyl-4,4-difluorocyclohexanecarboxylate
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Ethyl 4,4-difluorocyclohexanecarboxylate
- CAS Number:178312-47-5
- Formula:C₉H₁₄F₂O₂
- MDL Number:MFCD03788494
- Molecular Weight:192.21 g/mol
- Boiling Pt:84…86 °C/8mmHg
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: Ethyl 4,4-difluorocyclohexanecarboxylate
- CAS Number:178312-47-5
- Formula:C₉H₁₄F₂O₂
- MDL Number:MFCD03788494
- Molecular Weight:192.21 g/mol
- Boiling Pt:84…86 °C/8mmHg
- Storage Temperature:Ambient



