
(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:127852-28-2
:
Formula:C₁₀H₈F₆O
MDL Number:MFCD03093010
:258.16 g/mol
Melting Pt:53…58 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together