
Bis(trifluoromethanesulphonyl)imide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:82113-65-3
:
Formula:C₂HF₆NO₄S₂
MDL Number:MFCD00214154
:281.143 g/mol
Boiling Pt:90…91 °C
Melting Pt:46…57 °C
Density:1.36 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3261
ADR:8,I
:GHS05
Frequently Bought Together