
3,5-Bis(trifluoromethyl)phenylboronic acid 98%
:73852-19-4
:
:C₈H₅BF₆O₂
:MFCD00051850
:217…221 °C
:257.93 g/mol
:GHS07
:Ambient
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
4-Pentyn-1-ol ≥98%
: Thermo Fisher Scientific
Germanium(IV) chloride ≥99.9999% (metals basis)
: Thermo Fisher Scientific