
2-(4-Methoxyphenoxy)-5-(trifluoromethyl)aniline 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:62966-74-9
:
Formula:C₁₄H₁₂F₃NO₂
MDL Number:MFCD00792432
:283.25 g/mol
Melting Pt:73…75 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together