
4-Methoxy-3-(trifluoromethyl)benzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:261951-87-5
:
Formula:C₉H₆F₃NO
MDL Number:MFCD01631575
:201.15 g/mol
Melting Pt:98…100 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
Frequently Bought Together