
4-Fluoro-3-(trifluoromethyl)anisole 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:127271-65-2
:
Formula:C₈H₆F₄O
MDL Number:MFCD01631405
:194.13 g/mol
Flash Pt:67 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together