

3-Fluoro-5-(trifluoromethyl)anisole 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:261951-79-5
:
Formula:C₈H₆F₄O
MDL Number:MFCD01631491
:194.13 g/mol
Flash Pt:52 °C
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together