
4-(Trifluoromethylthio)benzyl alcohol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56456-52-1
:
Formula:C₈H₇F₃OS
MDL Number:MFCD00236342
:208.20 g/mol
Melting Pt:49…54 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together