
4-Fluoro-3-toluoyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:455-84-5
:
Formula:C₈H₆ClFO
MDL Number:MFCD01631426
:172.59 g/mol
Boiling Pt:129…133 °C/17mmHg
Density:1.215 g/cm³ (25 °C)
Flash Pt:98 °C
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together