
3-(2-Fluorophenyl)propionic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1643-26-1
:
Formula:C₉H₉FO₂
MDL Number:MFCD01310820
:168.17 g/mol
Melting Pt:74…79 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together