
2-(4-Fluorophenyl)malondialdehyde 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:493036-47-8
:
Formula:C₉H₇FO₂
MDL Number:MFCD02261897
:166.15 g/mol
Melting Pt:165…167 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together