
2,5-Difluoro-4-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:114973-22-7
:
Formula:C₇H₄F₅N
MDL Number:MFCD00190121
Boiling Pt:195…197 °C
Density:1.51 g/cm³ (25 °C)
UN:2810
ADR:6.1,III
:197.1054 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together