
3,3-Bis(trifluoromethyl)-3-hydroxypropionic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1547-36-0
:
Formula:C₅H₄F₆O₃
MDL Number:MFCD00040910
:226.08 g/mol
Melting Pt:84…85 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together