Order Entry
Czech Republic
ContactUsLinkComponent
2-((Difluoromethyl)thio)benzoic acid 97%
Irritant
2-((Difluoromethyl)thio)benzoic acid 97%
Catalog # APOSPC0270-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-((Difluoromethyl)thio)benzoic acid 97%
Catalog # APOSPC0270-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :204.2 g/mol
Melting Pt:150…152 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:79676-56-5
Formula:C₈H₆F₂O₂S