4-Chloro-3-(trifluoromethyl)anisole 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:400-73-7
:
Formula:C₈H₆ClF₃O
MDL Number:MFCD01631404
:210.58 g/mol
Boiling Pt:113 °C
Storage Temperature:Ambient
UN:3272
ADR:3,III
Frequently Bought Together