
3-Chloro-4-(trifluoromethoxy)benzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:129604-26-8
:
Formula:C₈H₃ClF₃NO
MDL Number:MFCD01631557
:221.57 g/mol
Melting Pt:39…41 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
Frequently Bought Together