
1-Methyl-1-propylpyrrolidinium bis(trifluoromethanesulphonyl)imide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:223437-05-6
:
Formula:C₁₀H₁₈F₆N₂O₄S₂
MDL Number:MFCD08458917
:408.37 g/mol
Melting Pt:12 °C
Density:1.44 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together